Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRp55 Mouse anti-Human, Clone: 9-1-56, MilliporeSigma™

Catalog No. MABE152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE152MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE152MI Supplier MilliporeSigma Supplier No. MABE152
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-SRp55 Antibody, clone 9-1-56 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry to detect SRp55 also known as arginine/serine-rich splicing factor 6, Pre-mRNA-splicing factor SRP55.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRp55
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9-1-56
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl without 0.05% sodium azide.
Gene Accession No. NP_006266
Gene Symbols SRP55, SFRS6, B52
Host Species Mouse
Immunogen MBP-tagged recombinant protein corresponding to human SRp55.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.