Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRp55 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 061366MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-136-6MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-136-6MI Supplier MilliporeSigma Supplier No. 061366

Rabbit Polyclonal Antibody

Anti-SRp55 Antibody is a Rabbit Polyclonal Antibody for detection of SRp55 also known as serine/arginine-rich splicing factor 6, Pre-mRNA-splicing factor SRP55 and has been validated in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRp55
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols SRP55; SFRS6; SRSF6; B52
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human SRp55.
Purification Method Protein A
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_006266
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.