Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRPK2 Mouse, Unlabeled, Clone: 23, BD

Catalog No. BDB611118 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611118 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611118 Supplier BD Biosciences Supplier No. 611118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mammalian cell pre-mRNA splicing is mediated by the spliceosome, a multi-component complex that contains two types of splicing factors: small nuclear ribonucleoprotein particles (snRNPs) and non-snRNP factors. Interactions between snRNPs and pre-mRNA ensures proper establishment of a catalytic core for the splicing reaction. However, these interactions are mediated by the non-snRNP factors. The superfamily of Arg/Ser-rich (RS) domain containing splicing factors are well known non-snRNPs. All of these proteins share a similar structure consisting of an N-terminal RNA recognition motif and a C-terminal RS domain. However, different SR factors have distinct specificities and their function is regulated by their level of expression and by reversible phosphorylation. Two families of kinases phosphorylate RS domain-containing proteins: SR protein-specific kinases (SRPK1 and 2) and Clk/Sty. SRPK1 is predominate in the pancreas, whereas SRPK2 is highly expressed in brain. SRPK2 contains an N-terminal proline-rich sequence that mediates its in vitro interaction with a WW domain containing protein. SRPKs affect splice-site selection and are thought to affect alternative splicing.

Host Species: Mouse
Clone: 23
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human SRPK2 aa. 363-485
Formula Weight [Chemical]: 115kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPK2
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Description SR Protein specific Kinase-2
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human SRPK2 aa. 363-485
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.