Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-1 Mouse, Alexa Fluor 555, Clone: MC480, BD

Catalog No. BDB560119 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560119 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560119 Supplier BD Biosciences Supplier No. 560119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MC480 monoclonal antibody reacts with Stage-Specific Embryonic Antigen-1 (SSEA-1), which is a terminal carbohydrate epitope (3-fucosyl-N-acetyllactosamine or 3-FAL) on glycoproteins and lactose series glycolipids. SSEA-1 is related to Lewis blood group antigens and is found in a variety of embryonic and adult tissues and cancers. As its name implies, the expression of SSEA-1 is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. However, its expression pattern differs in the human and mouse. In the human, SSEA-1 is not found on embryonic stem (ES) cells, embryonic inner cell mass (ICM), or teratocarcinoma (embryonal carcinoma or EC) cells. As human EC and ES cells undergo differentiation, SSEA-1 expression is upregulated. In the adult, the same epitope is expressed as CD15 on granulocytes and monocytes, but not lymphocytes or dendritic cells. In the mouse, SSEA-1 is found on EC, ES, and primordial germ cells, 8-cell to blastocyst embryos, ICM, and on subpopulations of cells in the adult central nervous system, including stem cells. In contrast to the human, SSEA-1 expression is reduced as mouse EC and ES cells undergo differentiation.

Host Species: Mouse
Clone: MC480
Isotype: IgM, κ
Species Reactivity: Human, Mouse
Immunogen: Mouse Teratocarcinoma Cell Line

Bioimaging

TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA-1
Applications In vitro Assay
Classification Monoclonal
Clone MC480
Conjugate Alexa Fluor 555
Description 3-FAL, X-hapten, LeX antigen, CD15
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Teratocarcinoma Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.