Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SSEA-1 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$233.50 - $514.50
Specifications
Antigen | SSEA-1 |
---|---|
Clone | MC-480 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2155C025
![]() |
R&D Systems
FAB2155C025 |
25 Tests |
Each for $233.50
|
|
|||||
FAB2155C100
![]() |
R&D Systems
FAB2155C100 |
100 Tests |
Each for $514.50
|
|
|||||
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
SSEA-1 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Mouse | |
Human, Mouse | |
alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA1, SSEA-1, stage-specific embryonic antigen 1 | |
FUT4 | |
IgM | |
Affinity Purified | |
Detects human and mouse SSEA-1. |
MC-480 | |
Flow Cytometry | |
PerCP | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
2526 | |
F9 mouse teratocarcinoma stem cells | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title