Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-3 Rat, Alexa Fluor 647, Clone: MC-631 (also known as MC631), BD

Catalog No. BDB561145 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561145 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561145 Supplier BD Biosciences Supplier No. 561145
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MC-631 monoclonal antibody reacts with Stage-Specific Embryonic Antigen-3 (SSEA-3), a carbohydrate epitope on the major ganglioside, but not the neutral glycolipid, of mouse embryos and human teratocarcinoma cells. As its name implies, the expression of SSEA-3 is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. However, its expression pattern differs in the human and mouse. In the human, SSEA-3 is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (ICM), and embryonic stem (ES) cells, and erythrocytes. As human stem cells undergo differentiation, SSEA-3 expression diminishes. In the mouse, SSEA-3 is found on oocytes, ova, zygotes, early cleavage-stage embryos, early blastocysts, ICM, primitive endoderm, and adult kidney, but not on EC or ES cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA-3
Applications Flow Cytometry
Classification Monoclonal
Clone MC-631 (also known as MC631)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Embryos
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.