Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

slingshot homolog 3 (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-506 Supplier Abnova Corporation Supplier No. H00054961D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SSH3 protein.

The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM

Sequence: MALVTVSRSPPGSGASTPVGPWDQAVQRRSRLQRRQSFAVLRGAVLGLQDGGDNDDAAEASSEPTEKAPSEEELHGDQTDFGQGSQSPQKQEEQRQHLHLMVQLLRPQDDIRLAAQLEAPRPPRLRYLLVVSTREGEGLSQDETVLLGVDFPDSSSPSCTLGLVLPLWSDTQVYLDGDGGFSVTSGGQSRIFKPISIQTMWATLQVLHQACEAALGSGLVPGGSALTWASHYQERLNSEQSCLNEWTAMADLESLRPPSAEPGGSSEQEQMEQAIRAELWKVLDVSDLESVTSKEIRQALELRLGLPLQQYRDFIDNQMLLLVAQRDRASRIFPHLYLGSEWNAANLEELQRNRVTHILNMAREIDNFYPERFTYHNVRLWDEESAQLLPHWKETHRFIEAARAQGTHVLVHCKMGVSRSAATVLAYAMKQYECSLEQALRHVQELRPIARPNPGFLRQLQIYQGILTASRQSHVWEQKVGGVSPEEHPAPEVSTPFPPLPPEPEGGGEEKVVGMEESQAAPKEEPGPRPRINLRGVMRSISLLEPSLELESTSETSDMPEVFSSHESSHEEPLQPFPQLARTKGGQQVDRGPQPALKSRQSVVTLQGSAVVANRTQAFQEQEQGQGQGQGEPCISSTPRFRKVVRQASVHDSGEEGEA

Specifications

Antigen slingshot homolog 3 (Drosophila)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SSH3 protein.
Formulation PBS with no preservative; pH 7.4
Gene SSH3
Gene Accession No. BC007709.1
Gene Alias FLJ10928/FLJ20515/SSH-3
Gene Symbols SSH3
Host Species Rabbit
Immunogen SSH3 (AAH07709.1, 1 a.a. ∼ 659 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 54961
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.