Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSPN Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535287
Description
Component of the dystrophin-glycoprotein complex (DGC), a complex that spans the muscle plasma membrane and forms a link between the F-actin cytoskeleton and the extracellular matrix. Preferentially associates with the sarcoglycan subcomplex of the DGC.
Specifications
| SSPN | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q14714 | |
| SSPN | |
| KLH conjugated synthetic peptide between 6-34 amino acids from the N-terminal region of human SSPN | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| SSPN | |
| SSPN;KRAG; Sarcospan; Sarcospan; K-ras oncogene-associated protein; Sarcospan; Kirsten-ras-associated protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8082 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction