Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST2A1 Mouse anti-Human, Mouse, Clone: 4A10.2, MilliporeSigma™

Catalog No. MABD32MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD32MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD32MI Supplier MilliporeSigma Supplier No. MABD32

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Anti-ST2A1 Antibody, clone 4A10.2 detects level of ST2A1 and has been published and validated for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ST2A1
Applications Western Blot
Classification Monoclonal
Clone 4A10.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_003158
Gene Symbols SULT2A1, HST, STD
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human ST2A1.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.