Learn More
ST3 beta-galactoside alpha-2,3-sialyltransferase 4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ST3GAL4.
Supplier: Abnova Corporation H00006484A01
Description
Synthesis of alpha-2,3-linked sialic acid to Gal(beta-1,3)GalNAc is mediated by at least 3 distinct beta-galactoside alpha-2,3-sialyltransferases (EC 2.4.99.4), including ST3GAL4. In contrast, only a single gene encodes the beta-galactoside alpha-2,6-sialyltransferase (EC 2.4.99.1), ST6GAL1 (MIM 109675) (Chang et al., 1995 [PubMed 7655169]).[supplied by OMIM
Sequence: FYFPIPEKKEPCLQGEAESKASKLFGNYSRDQPIFLRLEDYFWVKTPSAYELPYGTKGSEDLLLRVLAITSSSIPKNIQSLRCRRCVVVGNGHRLRNSSLSpecifications
ST3 beta-galactoside alpha-2,3-sialyltransferase 4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ST3GAL4. | |
ST3GAL4 | |
CGS23/FLJ11867/FLJ46764/NANTA3/SAT3/SIAT4/SIAT4C/ST3GalIV/STZ | |
Mouse | |
50 μL | |
Yes | |
6484 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006278 | |
ST3GAL4 | |
ST3GAL4 (NP_006269, 31 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.