Learn More
ST6GAL1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ST6GAL1 peptide using ARM Technology.
Supplier: Abnova Corporation H00006480K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of glycosyltransferase family 29. The encoded protein is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The protein, which is normally found in the Golgi but can be proteolytically processed to a soluble form, is involved in the generation of the cell-surface carbohydrate determinants and differentiation antigens HB-6, CD75, and CD76. This gene has been incorrectly referred to as CD75. Three transcript variants encoding two different isoforms have been described. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ST6GAL1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ST6GAL1 | |
ST6GAL1 | |
A synthetic peptide of human ST6GAL1 | |
RUO | |
6480 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ST6 beta-galactosamide alpha-2,6-sialyltranferase 1 | |
ST6GAL1 | |
CD75, MGC48859, SIAT1, ST6GalI, ST6N | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.