Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-586 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-586 Supplier Abnova Corporation Supplier No. H00008128D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ST8SIA2 protein.

The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29. [provided by RefSeq

Sequence: MQLQFRSWMLAALTLLVVFLIFADISEIEEEIGAEVVINGSSSPAVVDRSNESIKHNIQPASSKWRHNQTLSLRIRKQILKFSDAEKDISVLKGTLKPGDIIHYIFDRDSTMNVSQNLYELLPRTSPLKNKHFGTCAIVGNSGVLLNSGCGQEIDAHSFVIRCNLAPVQEYARDVGLKTDLVTMNPSVIQRAFEDLVNATWREKLLQRLHSLNGSILWIPAFMARGGKERVEWVNELILKHHVNVRTAYPSLRLLHAVRGYWLTNKVHIKRPTTGLLMYTLATRFCKQIYLYGFWPFPLDQNQNPVKYHYYDSLKYGYTSQASPHTMPLEFKALKSLHEQGALKLTVGQCDGAT

Specifications

Antigen ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ST8SIA2 protein.
Formulation PBS with no preservative; pH 7.4
Gene ST8SIA2
Gene Accession No. BC096205
Gene Alias HsT19690/MGC116854/MGC116857/SIAT8B/ST8SIA-II/STX
Gene Symbols ST8SIA2
Host Species Rabbit
Immunogen ST8SIA2 (AAH96205.1, 1 a.a. ∼ 354 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neurobiology
Primary or Secondary Primary
Gene ID (Entrez) 8128
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.