Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stage-Specific Embryonic Antigen-1 Mouse anti-Human, Mouse, Rat, AlexaFluor™488, Clone: MC-480, MilliporeSigma™

Catalog No. MAB4301XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4301XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4301XMI Supplier MilliporeSigma Supplier No. MAB4301X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, Alexa Fluor 488 is validated for use in flow cytometry, immunocytochemistry, immunofluorescence for the detection of Stage-Specific Embryonic Antigen-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stage-Specific Embryonic Antigen-1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MC-480
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin conjugated to Alexa Fluor™ 488. Liquid in Phosphate buffer with 15mg/mL BSA as a stabilizer and 0.1% sodium azide
Gene Accession No. NM_002033.2
Gene Symbols FUT4, ELFT, FCT3A, Fuc-TIV, 1,3fucosyltransferase, FUC-TIV, FucT-IV, CD15, Alpha-, EC 2.4.1.-
Host Species Mouse
Immunogen F9 teratocarcinoma stem cells (X-irradiated)
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 24 months. Protect from Light.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.