Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Stage-Specific Embryonic Antigen-1 Mouse anti-Human, Mouse, Rat, Clone: MC-480

Catalog No. MAB4301MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4301MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4301MI Supplier MilliporeSigma™ Supplier No. MAB4301
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480 detects level of Stage-Specific Embryonic Antigen-1 and has been published and validated for use in flow cytometry, immunofluoresence, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stage-Specific Embryonic Antigen-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone MC-480
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgM in buffer containing 0.05M Potassium Phosphate, 0.3M NaCl, pH 8.0 with 0.05% Sodium Azide
Gene Accession No. NM_002033.2
Gene Symbols FUT4, ELFT, FCT3A, Fuc-TIV, 1,3fucosyltransferase, FUC-TIV, FucT-IV, CD15, Alpha-, EC 2.4.1.-
Host Species Mouse
Immunogen F9 tetracarcinoma stem cells (X-irradiated).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.