Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanniocalcin-1 Rabbit anti-Human, Clone: EPR6055, MilliporeSigma™

Catalog No. MABC738MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC738MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC738MI Supplier MilliporeSigma Supplier No. N

Rabbit Monoclonal Antibody

This Anti-Stanniocalcin-1, clone EPR6055, Rabbit Monoclonal Antibody is validated for use in Western Blotting for the detection of Stanniocalcin-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stanniocalcin-1
Applications Western Blot
Classification Monoclonal
Clone EPR6055
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human Stanniocalcin-1.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) STC1, STC
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.