Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus aureus Mouse anti-Human, Clone: STAPH 11-248.2, MilliporeSigma™

Catalog No. MAB930MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB930MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB930MI Supplier MilliporeSigma Supplier No. MAB930
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Staphylococcus aureus using this Anti-Staphylococcus aureus Antibody, clone STAPH 11-248.2 validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus aureus
Applications ELISA
Classification Monoclonal
Clone STAPH 11-248.2
Conjugate Unconjugated
Formulation Ascites fluid. Liquid containing no preservatives.
Host Species Mouse
Immunogen UV-inactivated Staphylococcus aureas cells (ATCC# 29740).
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.