Learn More
STARD7, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human STARD7 peptide using ARM Technology.
Supplier: Abnova Corporation H00056910K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Although the function of this gene is not known, its existence is supported by mRNA and EST data. The predicted gene product contains a region similar to the STAR-related lipid transfer (START) domain, which is often present in proteins involved in the cell signaling mediated by lipid binding. Alternatively spliced transcript variants have been described, although some transcripts occur only in cancer cell lines. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
STARD7 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
STARD7 | |
STARD7 | |
A synthetic peptide of human STARD7 | |
RUO | |
56910 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
StAR-related lipid transfer (START) domain containing 7 | |
STARD7 | |
GTT1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.