Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat1 Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB610185 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610185 50 μg
BDB610186 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610185 Supplier BD Biosciences Supplier No. 610185
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Stat proteins function as both cytoplasmic signal transducers and activators of transcription. The Stat91/84 (the two proteins are the result of alternate splicing-Stat91 has an additional 38 C-terminal amino acids) and Stat113 were the first identified members of this protein family. These three polypeptides contain both SH2 and SH3 domains and have also been described as members of the ISGF3 (interferon-stimulated gene factor 3) complex. With the discovery of additional members of the Stat family (Stats3 & 4), the nomenclature has been revised to indicate the Stat family members in the order of their discovery. Stat 91, 84, and 113 have become Stat1α, Stat1β, and Stat2, respectively. Stat1α is present in a higher concentration than Stat1β in most cell types. In response to IFNα treatment, Stat1α, Stat1β, and Stat2 become tyrosine-phosphorylated and migrate to the nucleus where they join a 48kDa DNA binding protein and subsequently direct the transcription at IFNα responsive elements. In IFN-γ treated cells, Stat1α (but not Stat2) becomes phosphorylated and forms a dimer. It then enters the nucleus and binds to the IFN-γ activated site (GAS) element in order to direct IFN-γ activated transcription.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat1
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Description C-Terminus
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.