Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06501
Description
Detect STAT1 also known as Signal Transducer and Activator of Transcription 1 with Anti-STAT1 Antibody, CT (Rabbit Polyclonal Antibody), that has been demonstrated to work in EMSA, immunoprecipitation and western blotting.
Specifications
STAT1 | |
Polyclonal | |
Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl and 0.05% sodium azide, pH 7.4. | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Mouse | |
Purified |
Electromobility Shift Assay, Immunoprecipitation, Western Blot | |
Unconjugated | |
STAT1; ISGF-3; STAT91; DKFZp686B04100 | |
GST fusion-protein containing the carboxy terminal 36 amino acids of human STAT1α | |
250 μg | |
NM_007315 | |
2-8°C for 12 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction