Learn More
Description
Host Species: Mouse
Clone 4a
Isotype IgG2a, κ
Species Reactivity Hu, Ms, Rt
Immunogen Phosphorylated Human Stat1 Peptide
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Antigen | Stat1 (pY701) |
| Classification | Monoclonal |
| Clone | 4a |
| Host Species | Mouse |
| Immunogen | Phosphorylated Human Stat1 Peptide |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human, Mouse, Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.