Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT2, p Tyr690 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11086975
Description
STAT2 Polyclonal specifically detects STAT2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| STAT2 (p Tyr690) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| Rabbit | |
| 97.92 kDa | |
| 0.1 mg | |
| Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6773 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.15 mg/ml | |
| Western Blot 1:500-1:1000, ELISA 1:85000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| P52630 | |
| STAT2 | |
| This affinity purified STAT2 [p Tyr690] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a C-terminus region near phospho Tyrosine 690 of human STAT2 protein. (Uniprot: P52630) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This affinity purified antibody is directed against human STAT2 pY690 protein. A BLAST analysis was used to suggest cross-reactivity with STAT2 pY690 protein from human, rat (73%) and mouse (76%), sources based on homology with the immunizing sequence. Reactivity against homologues from other sources is not known. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction