Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat2 (pY690) Mouse anti-Human, Unlabeled, Clone: 7A/STAT2 (PY690), BD

Catalog No. BDB558095 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558095 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558095 Supplier BD Biosciences Supplier No. 558095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat1 and Stat2 are components of the ISGF3 (Interferon-Stimulated Gene Factor 3) complex, which is the primary transcription activator induced by the binding of the interferon to a specific cell-surface receptor. Stat2 is a 113kDa protein having approximately 40% homology with Stat1a. Stat2 interacts with Stat1 for the efficient activation of Stat1 in response to IFN-α, which is essential for normal transcriptional activity of the ISGF3 complex. Stat2 phosphorylation at the tyrosine 690 residue (Y690) after IFN-α activation can occur independently of Stat1, but the localization and nuclear stability of phosphorylated Stat2 is dependent on Stat1. The 7a/Stat2 monoclonal antibody recognizes the phosphorylated Y690 of human Stat2.

Host Species: Mouse
Clone: 7a
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human Stat2 Peptide

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat2 (pY690)
Applications Western Blot
Classification Monoclonal
Clone 7A/STAT2 (PY690)
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Stat2 Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.