Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT3 Mouse anti-Human, Mouse, Clone: 18F7.1, MilliporeSigma™

Catalog No. 051078MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-107-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-107-8MI Supplier MilliporeSigma Supplier No. 051078
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-STAT3 Antibody, clone 18F7.1 (mouse monoclonal antibody) validated in western blot, immunocytochemistry to detect STAT3 also known as Acute-phase response factor, DNA-binding protein APRF, signal transducer and activator of transcription 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 18F7.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_003141
Gene Symbols APRF, FLJ20882, HIES, MGC16063
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human STAT3.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.