Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

signal transducer and activator of transcription 3 (acute-phase response factor), Mouse, Clone: 4D6, Abnova™

Catalog No. 89000984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-984 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-984 Supplier Abnova Corporation Supplier No. H00006774M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant STAT3.

The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated through phosphorylation in response to various cytokines and growth factors including IFNs, EGF, IL5, IL6, HGF, LIF and BMP2. This protein mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of this protein. PIAS3 protein is a specific inhibitor of this protein. Three alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq

Sequence: LVYLYPDIPKEEAFGKYCRPESQEHPEADPGAAPYLKTKFICVTPTTCSNTIDLPMSPRTLDSLMQFGNNGEGAEPSAGGQFESLTFDMELTSECATSPM

Specifications

Antigen signal transducer and activator of transcription 3 (acute-phase response factor)
Applications ELISA, Immunofluorescence, In situ PLA, Western Blot
Classification Monoclonal
Clone 4D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant STAT3.
Formulation PBS with no preservative; pH 7.4
Gene STAT3
Gene Accession No. NM_003150
Gene Alias APRF/FLJ20882/HIES/MGC16063
Gene Symbols STAT3
Host Species Mouse
Immunogen STAT3 (NP_003141, 670 a.a. ∼ 769 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Receptors
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6774
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.