Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat3 Mouse, Unlabeled, Clone: 84, BD

Catalog No. BDB610189 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610189 50 μg
BDB610190 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610189 Supplier BD Biosciences Supplier No. 610189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Stat proteins function as both cytoplasmic signal transducers and activators of transcription. Stat91/84 (the two proteins are the result of alternate splicing-Stat91has an additional 38 C-terminal amino acids) and Stat113 were the first identified members of this protein family. With the discovery of additional members of the Stat family (Stats3 & 4), the nomenclature has been revised to indicate the Stat family members in the order of their discovery. Stat 91, 84, and 113 have become Stat1α, Stat1β, and Stat2, respectively. Stat3 is a 92kDa protein that is activated as a DNA binding protein through tyrosine phosphorylation in response to treatment of cells with EGF and IL-6 but not with IFN-γ. Stat3 is widely expressed and can bind to DNA in the absence of Stat1α or Stat2. Stat3 can bind to the sis-inducible element (SIE) site from the c-fos promoter. The site is similar to the GAS element that is present in IFN-γ induced genes. It appears that Stat3 binds to DNA as a homodimer, but it is also capable of binding as a heterodimer with Stat1. With the isolation of Stat3 and the discovery of Stat4, it appears that the Stat family may contain many members, each with a characteristic tissue distribution and specific activating ligands.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat3
Applications Western Blot
Classification Monoclonal
Clone 84
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.