Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat STAT3 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC1799F
This item is not returnable.
View return policy
Description
STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| STAT3 | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6774 | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| 232209 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P40763 | |
| STAT3 | |
| E. coli-derived recombinant human STAT3 Met1-Asn175 Accession # P40763 | |
| 100 Tests | |
| Primary | |
| The unconjugated antibody (Catalog # MAB1799) detects human, mouse, and rat STAT3 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction