Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat3 Rabbit anti-Human, Mouse, Rat, Bovine, Pig, Primate, Polyclonal, MilliporeSigma™

Catalog No. 072173MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-217-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-217-3MI Supplier MilliporeSigma Supplier No. 072173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Stat3 Antibody (rabbit polyclonal antibody) validated in western blot, immunoprecipitation to detect Stat3 also known as Acute-phase response factor, DNA-binding protein APRF, signal transducer and activator of transcription 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols APRF; HIES; STAT3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human STAT3 near the C-terminus.
Purification Method Protein A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_644805
Target Species Bovine, Human, Mouse, Pig, Primate, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.