Learn More
Description
Specifications
Specifications
| Antigen | STAT3 |
| Applications | Electromobility Shift Assay, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Protein A purified rabbit IgG in 200 L of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide. Frozen at −20°C. |
| Gene Symbols | APRF; FLJ20882; HIES; MGC16063 |
| Host Species | Rabbit |
| Immunogen | Bacterially expressed GST fusion protein corresponding to a.a. 688-722 of human STAT 3 (RPESQEHPEADPGSAAPYLKTKFICVTPTTCSNTI). The immunizing sequence is identical in mouse and rat STAT3. The first 28 amino acids are identical to mouse STAT3B. |
| Purification Method | Protein A purfied |
| Quantity | 200 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.