Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat3 (pS727) Mouse, Unlabeled, Clone: 49/P-STAT3, BD

Catalog No. BDB612543 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612543 150 μg
BDB612542 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612543 Supplier BD Biosciences Supplier No. 612543
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. Seven mammalian Stat proteins have been identified: Stat1-4, Stat5a, 5b, and Stat6. Stat3 is a 92kDa protein that is activated as a DNA binding protein through cytokines, such as IL-6, and growth factors, such as EGF. Stat3 is phophorylated at serine 727 (S727) via the MAPK pathway. The S727 residue is located at a conserved Pro-X-Ser-Pro sequence, which is recognized by the protein kinase ERK. Activation through the S727 residue is thought to lead to initiation of transcription. Upon activation, Stat3 dimerizes, translocates to the nucleus, and binds DNA response elements thereby regulating gene expression. It appears that Stat3 binds to DNA as a homodimer, but it is also capable of binding as a heterodimer with Stat1. In addition to serine phosphorylation, Stat3 is also phosphorylated at tyrosine 705 by JAK1 in response to cytokine stimulation. Stat3 is widely expressed and can bind to the sis-inducible element (SIE) site from the c-fos promoter. This site is similar to the GAS element that is present in IFN-g-induced genes. Thus, phosphorylation of S727 in Stat3 occurs in response to growth factors and cytokines, and is essential for normal transcription activity.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Stat3 (pS727)
Applications Western Blot
Classification Monoclonal
Clone 49/P-STAT3
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Stat3
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.