Learn More
Description
Host Species: Mouse
Clone 47/Stat5(pY694)
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Phosphorylated Sheep Stat5 Peptide
Intracellular block/Flow Cytometry
Specifications
Specifications
| Antigen | Stat5 (pY694) |
| Classification | Monoclonal |
| Clone | 47/Stat5(pY694) |
| Conjugate | Brilliant Violet 421 |
| Host Species | Mouse |
| Immunogen | Phosphorylated Sheep Stat5 Peptide |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.