Learn More
STAT5A/B Rabbit anti-Human, Mouse, Rat, Sheep, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | STAT5A/B |
| Applications | Electromobility Shift Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | whole rabbit antiserum containing 0.05% sodium azide |
| Gene Symbols | STAT5A; MGF; STAT5 |
| Host Species | Rabbit |
| Immunogen | peptide (CQELQKKAEHQVGEDG), corresponding to amino acids 66-80 of human STAT5 A and B |
| Purification Method | Antiserum |
| Quantity | 250 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.