Learn More
signal transducer and activator of transcription 5A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of this protein in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of this gene is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this gene in cells. [provided by RefSeq
Specifications
Specifications
| Antigen | signal transducer and activator of transcription 5A |
| Applications | In situ PLA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human STAT5A protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | STAT5A |
| Gene Accession No. | NM_003152.2 |
| Gene Alias | MGF/STAT5 |
| Gene Symbols | STAT5A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.