Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5B Rabbit anti-Avian, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06969MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-969-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-969-MI Supplier MilliporeSigma Supplier No. 06969

Rabbit Polyclonal Antibody

Detect STAT5B also known as signal transducer and activator of transcription 5B with Anti-STAT5B Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT5B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation rabbit antiserum
Gene Symbols STAT5B; STAT5
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (MDSQWIPHAQS) corresponding to amino acids 776-786 of mouse STAT5B
Purification Method Antiserum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_012448
Target Species Avian, Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.