Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Stathmin, Polyclonal

Catalog No. 569391100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
56-939-1100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-939-1100UL Supplier MilliporeSigma™ Supplier No. 569391100UL

Rabbit Polyclonal Antibody

Specifically detects Stathmin Clone: in samples, and it is validated for Immunohistochemistry, Immunoprecipitation

Stathmin is a highly conserved cytoplasmic phosphoprotein, which is thought to act as a relay that integrates diverse cell signaling pathways. Stathmin interferes with the dynamic instability of microtubulesin vitro andin vivo. Stathmin in vitro either promotes rapid depolymerization of microtubules or prevents microtubule assembly in a polymerization inhibition assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stathmin
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids near the C-terminus of mammalian stathmin
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽17kDa stathmin protein.
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.