Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staufen Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5781MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5781MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5781MI Supplier MilliporeSigma Supplier No. AB5781

Rabbit Polyclonal Antibody

Anti-Staufen Antibody is an antibody against Staufen for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staufen
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.
Gene Symbols STAU1; FLJ25010; STAU; staufen
Host Species Rabbit
Immunogen Synthetic peptide from amino acids 458-472 of human Staufen 1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001037328.1; NM_004602.2; NM_017452.2; NM_017453.2; NM_017454.2
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.