Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STEP Mouse anti-Human, Mouse, Rat, Clone: 23E5, MilliporeSigma™

Catalog No. 05730MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-730-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-730-MI Supplier MilliporeSigma Supplier No. 05730
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-STEP Antibody, clone 23E5 is an antibody against STEP for use in immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEP
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23E5.
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_019253
Gene Symbols PTPN5, FLJ14427, STEP, PTPSTEP
Host Species Mouse
Immunogen Peptide corresponding to amino acids 57-74 of rat STEP46
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.