Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ STING, Rat anti-Mouse, Clone: 41

Catalog No. MABF213MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF213MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF213MI Supplier MilliporeSigma™ Supplier No. MABF213
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti- STING, clone 41 Antibody is validated for use in Western Blotting and Flow Cytometry for the detection of STING.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STING
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 41
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_082537
Gene Symbols Tmem173, Eris Mita, Mpys, Sting
Host Species Rat
Immunogen Recombinant protein corresponding to the cytoplasmic domain of mouse STING.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.