Learn More
STING Mouse anti-Human, Unlabeled, Clone: T3-680, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564835
Description
The T3-680 monoclonal antibody specifically recognizes human STING (Stimulator of interferon genes protein) which is also known as Endoplasmic reticulum interferon stimulator (ERIS) and Mediator of IRF3 activation (MITA). STING is encoded by TMEM173 (Transmembrane protein 173). STING is a multi-pass transmembrane adaptor protein which is expressed in the endoplasmic reticulum of cells. STING acts as a sensor of cytosolic double-stranded DNA derived from bacteria and viruses and promotes the production of type I interferons, IFN- α and IFN- β . Following activation, STING translocates with TBK1kinase to perinuclear endosomes. The TBK1 kinase phosphorylates and activates interferon regulatory factors, such as IRF3, and NF-κB, which leads to the induction of type I interferon and other immune response genes that play roles in innate and adaptive immunity.

Specifications
STING | |
Monoclonal | |
0.5mg/mL | |
hSTING; MITA; hMITA; ERIS; MPYS; NET23; TM173; TMEM173 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
T3-680 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human STING Recombinant Protein | |
0.1 mg | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.