Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serine/threonine kinase 19, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016956 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-956 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-956 Supplier Abnova Corporation Supplier No. H00008859B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human STK19 protein.

This gene encodes a serine/threonine kinase which localizes predominantly to the nucleus. Its specific function is unknown; it is possible that phosphorylation of this protein is involved in transcriptional regulation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6 and expresses two transcript variants. [provided by RefSeq

Sequence: MQKWFSAFDDAIIQRQWRANPSRGGGGVSFTKEVDTNVATGAPPRRQRVPGRACPWREPIRGRRGARPGGGDAGGTPGETVRHCSAPEDPIFRFSSLHSYPFPGTIKSRDMSWKRHHLIPETFGVKRRRKRGPVESDPLRGEPGSARAAVSELMQLFPRGLFEDALPPIVLRSQVYSLVPDRTVADRQLKELQEQGEIRIVQLGFDLDAHGIIFTEDYRTRVLKACDGRPYAGAVQKFLASVLPACGDLSFQQDQMTQTFGFRDSEITHLVNAGVLTVRDAGSWWLAVPGAGRFIKYFVKGRQAVLSMVRKAKYRELLLSELLGRRAPVVVRLGLTYHVHDLIGAQLVDCISTTSGTLLRLPET

Specifications

Antigen serine/threonine kinase 19
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human STK19 protein.
Formulation PBS with no preservative; pH 7.4
Gene STK19
Gene Accession No. NM_004197.1
Gene Alias D6S60/D6S60E/G11/HLA-RP1/MGC117388/RP1
Gene Symbols STK19
Host Species Mouse
Immunogen STK19 (NP_004188.1, 1 a.a. ∼ 364 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 8859
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.