Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serine/threonine kinase 25 (STE20 homolog, yeast), Mouse, Clone: 4B10, Abnova™

Catalog No. 89001181 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-181 Supplier Abnova Corporation Supplier No. H00010494M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant STK25.

Sequence: FPPTIRPSPHSKLHKGTALHSSQKPAEPVKRQPRSQCLSTLVRPVFGELKEKHKQSGGSVGALEELENAFSLAEESCPGISDKLMVHLVERVQRFSHNRNHLTSTR

Specifications

Antigen serine/threonine kinase 25 (STE20 homolog, yeast)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant STK25.
Formulation PBS with no preservative; pH 7.4
Gene STK25
Gene Accession No. BC007852
Gene Alias DKFZp686J1430/SOK1/YSK1
Gene Symbols STK25
Host Species Mouse
Immunogen STK25 (AAH07852, 321 a.a. ∼ 426 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10494
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.