Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serine/threonine kinase 32C, Mouse, Clone: 4D12, Abnova™

Catalog No. 89001411 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-411 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-411 Supplier Abnova Corporation Supplier No. H00282974M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant STK32C.

The protein encoded by this gene is a member of the serine/threonine protein kinase family. The specific function of this kinase is not known. [provided by RefSeq

Sequence: MYAMKYMNKQQCIERDEVRNVFRELEILQEIEHVFLVNLWYSFQDEEDMFMVVDLLLGGDLRYHLQQNVQFSEDTVRLYICEMALALDYLRGQHIIHRDVKPDNILLDERGHAHLTDFNIATIIKDGERATALAGTKPYMAPEIFHSFVNGGTGYSFEVDWWSVGVMAYELLRGWRPYDIHSSNAVESLVQLFSTVSVQYVPTWSKEMVALLRKLLTVNPEHRLSSLQDVQAAPALAGVLWDHLSEKRVEPGFVPNKGRLHCDPTFELEEMILESRPLHKKKKRLAKNKSRDNSRDSSQSENDYLQDCLDAIQQDFVIFNREKLKRSQDLPREPLPAPESRDAAEPVEDEAERSALPMCGPICPSAGSG

Specifications

Antigen serine/threonine kinase 32C
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant STK32C.
Formulation PBS with no preservative; pH 7.4
Gene STK32C
Gene Accession No. BC015792
Gene Alias MGC23665/PKE/RP11-140A10.1/YANK3
Gene Symbols STK32C
Host Species Mouse
Immunogen STK32C (AAH15792, 1 a.a. ∼ 369 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 282974
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.