Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STMN2, Mouse, Clone: 3B12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant STMN2.
Supplier: Abnova Corporation H00011075M01A
Description
Superior cervical ganglion-10 is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin (MIM 151442).[supplied by OMIM
Sequence: MAKTAMAYKEKMKELSMLSLICSCFYPEPRNINIYTYDDMEVKQINKRASGQAFELILKPPSPISEAPRTLASPKKKDLSLEEIQKKLEASpecifications
STMN2 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_007029 | |
STMN2 | |
STMN2 (NP_008960, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
11075 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3B12 | |
Mouse monoclonal antibody raised against a partial recombinant STMN2. | |
STMN2 | |
SCG10/SCGN10/SGC10 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction