Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STMN3, Mouse, Clone: 1F2, Abnova™

Catalog No. 89123132 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-132 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-132 Supplier Abnova Corporation Supplier No. H00050861M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant STMN3.

The protein encoded by this gene belongs to the stathmin/oncoprotein 18 family of microtubule-destabilizing phosphoproteins. It is similar to the SCG10 protein and is involved in signal transduction and regulation of microtubule dynamics. [provided by RefSeq

Sequence: MASTISAYKEKMKELSVLSLICSCFYTQPHPNTVYQYGDMEVKQLDKRASGQSFEVILKSPSDLSPESPMLSSPPKKKDTSLEELQKRLEAAEERRKTQEAQVLKQLAERREHEREVLHKALEENNNFSRQAEEKLNYKMELSKEIREAHLAALRERLREKELHAAEVRRNKEQREEITG

Specifications

Antigen STMN3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant STMN3.
Formulation ascites with no preservative
Gene STMN3
Gene Accession No. BC009381
Gene Alias SCLIP
Gene Symbols STMN3
Host Species Mouse
Immunogen STMN3 (AAH09381, 1 a.a. ∼ 180 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50861
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.