Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human STRA6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5904
This item is not returnable.
View return policy
Description
STRA6 Monoclonal specifically detects STRA6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
STRA6 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
AAH25256 | |
STRA6 | |
NS0 mouse myeloma cell line transfected with human STRA6 Met1-Pro667 Accession # AAH25256 | |
100 μg | |
Primary | |
Detects human STRA6 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
496613 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ12541, MCOPS9, stimulated by retinoic acid gene 6 homolog (mouse), stimulated by retinoic acid gene 6 protein homolog | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64220 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction