Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRAP Mouse, Unlabeled, Clone: 22, BD

Catalog No. BDB611346 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611346 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611346 Supplier BD Biosciences Supplier No. 611346
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The transforming growth factor β (TGF-β) family of cytokines plays diverse and important roles in growth, development, and differentiation. Three high affinity TGF-β receptor types have been characterized. Both type I and type II receptors are type I transmembrane proteins which contain a Ser/Thr-kinase in their cytosolic domains. Type III receptor, also known as betaglycan, has no cytosolic signaling motif and functions in the presentation of TGF-β to the type I and type II receptors. The cytosolic domain kinase of the type I receptor is thought to be transphosphorylated by the corresponding kinase of the type II receptor, leading to activation of downstream substrates. Ser/Thr receptor associated protein (STRAP) contains six WD domains, which are characteristic of signal transduction proteins. STRAP can associate with both TGF-βI and TGF-βII receptors and overexpression of STRAP leads to inhibition of TGF-β-dependent transcriptional activation. STRAP is expressed ubiquitously in species ranging from yeast to human. Thus, STRAP may be an important element of TGF-β signal transduction pathways in a variety of tissues and species.

Host Species: Mouse
Clone: 22
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse STRAP aa. 42-154
Formula Weight [Chemical]: 39kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen STRAP
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse STRAP aa. 42-154
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.