Learn More
Description
- Flow Cytometry
- Volume per test: 20μL
- Source: prepared chromatographically from Streptomyces avidinii
- Stabilizer: Gelatin
- Preservative: 0.1% Sodium azide
Specifications
Specifications
| Applications | Flow Cytometry |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.