Learn More
Description
Host Species:
Clone: Streptavidin
Species Reactivity: Ms
Immunogen: Streptavidin from Streptomyces avidinii
Flow Cytometry
Specifications
Specifications
| Antigen | Streptavidin |
| Classification | Monoclonal |
| Clone | Streptavidin |
| Concentration | 0.1mg/mL |
| Conjugate | Brilliant Violet 786 |
| Immunogen | Streptavidin from Streptomyces avidinii |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.