Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Pharmingen™ PE Streptavidin

Catalog No. bdb554061 Shop All BD Biosciences Products
Click to view available options
Quantity:
0.5 mg

SAv-PE is a useful second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis.

  • SAv-PE is a useful second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis.
  • Excitation at 488-nm light leads to a fluorescence emission maximum of ∼575 nm.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
Conjugate PE (Phycoerythrin)
Quantity 0.5 mg
Regulatory Status For research use only (RUO)
Content And Storage Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.