Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striatin Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB610838 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610838 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610838 Supplier BD Biosciences Supplier No. 610838
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Signal transmission between neurons is regulated by the Ca2+-dependent release of neurotransmitters at the synapse. Striatin is a protein
identified in the synaptosomal fraction of rat brain. Reports of sequence homology searches have shown that striatin belongs to a family of
proteins with the repeated WD motif (the β-transducin repeat or the WD-40 repeat) found in regulatory proteins. Striatin also binds
calmodulin in a Ca2+-dependent manner. Immunolocalization and mRNA expression found striatin in the central nervous system, particularly
enriched in neurons of the striatum and motor neurons. Furthermore, subcellular localization studies indicated that Striatin is tightly associated
with the plasma membrane. Thus, striatin may play an important role in the Ca2+-dependent events occurring at the synapse.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Striatin
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Striatin aa. 450-600
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.