Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stromal Cell-Derived Factor-1α Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1868PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1868PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1868PMI Supplier MilliporeSigma Supplier No. AB1868P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Stromal Cell-Derived Factor-1α using this Anti-Stromal Cell-Derived Factor-1α Antibody validated for use in ELISA, western blot, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stromal Cell-Derived Factor-1α
Applications ELISA, Neutralization, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Lyophilized from 0.5X PBS pH 7.2. Reconstitute with sterile distilled water.
Gene Symbols CXCL12; SDF1A; TPAR1; TLSF; SDF-1; SDF1; SDF1B; SCYB12; hIRH; TLSF-b; SDF-1b; PBSF; TLSF-a; SDF-1a
Host Species Rabbit
Immunogen Highly pure (≥98%) recombinant human SDF-1alpha.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000609.4; NM_001033886.1; NM_199168.2
Target Species Human
Content And Storage Lyophilized: −20°C for up to 12 months. Reconstituted: 2-4°C in undiluted aliquots for up to 6 weeks
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.